Skip to Content
Merck
vibrant-m

Fluorescein-5-isothiocyanate (FITC)

vibrant-m
Empirical Formula (Hill Notation):
C21H11NO5S
CAS Number:
Molecular Weight:
389.38

Products

Brand
Formula Weight
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing