
(+)-Garcinia acid

Synonym(s):
(−)-Hydroxycitric acid lactone, (2S,3S)-3-Hydroxy-5-oxotetrahydrofuran-2,3-dicarboxylic acid, Garcinia lactone
Empirical Formula (Hill Notation):
C6H6O7
CAS Number:
Molecular Weight:
190.11
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1