
1H,1H,2H,2H-Perfluorooctyl acrylate

Synonym(s):
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate, 1H,1H,2H,2H-perfluorooctyl acrylate, TFOA
Linear Formula:
H2C=CHCO2CH2CH2(CF2)5CF3
CAS Number:
Molecular Weight:
418.15
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing