HPLC Separation of an Oligonucleotide and Its Degradation Fragments on Proteomix® SAX-NP5
![HPLC Separation of an Oligonucleotide and Its Degradation Fragments on Proteomix® SAX-NP5 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/232/576/8b0ddd5a-94f0-4999-a71e-f2efeda302e5/800/8b0ddd5a-94f0-4999-a71e-f2efeda302e5.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 μm particles (Z777228)
mobile phase
(A) 25 mM Tris, 1 mM EDTA 10% acetonitrile (8.0 pH); (B) A + 1.0 M NaCl
gradient
0 to 75% B in 50 min
flow rate
0.5 mL/min
detector
sample
Oligonucleotide (MW 12196) and its degradation fragments
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies