Merck
  • Spatiotemporal transcriptome provides insights into early fruit development of tomato (Solanum lycopersicum).
Spatiotemporal transcriptome provides insights into early fruit development of tomato (Solanum lycopersicum).
Scientific reports (2016-03-19)
Shuaibin Zhang, Meng Xu, Zhengkun Qiu, Ketao Wang, Yongchen Du, Lianfeng Gu, Xia Cui
ABSTRACT
Early fruit development is crucial for crop production in tomato. After fertilization, the ovary undergoes cell division and cell expansion before maturation. Although the roles of regulatory signals such as hormone and carbohydrate during early fruit development have been studied, the spatial distribution and the sequential initiation of these regulatory signals still need to be explored. Using the tomato cultivar 'Moneymaker', we analyzed the transcriptome of the ovule and the ovary wall/pericarp dissected from four different stages of the early developing fruits by stereoscope. These datasets give us the whole picture about the spatial and temporal signal distribution in early development of ovule and pericarp. Our results indicate that the hormone signal was initiated in both ovule and pericarp after fertilization. After that, different signals were activated in ovule and pericarp due to their distinct developmental processes. Our study provides spatiotemporal regulatory landscape of gene expression with sequential information which was not studied by previous work and further strengthens the comprehension of the regulatory and metabolic events controlling early fruit development.
Sigma-Aldrich
Paraplast Plus®, for tissue embedding
Pricing and availability is not currently available.