Merck
  • Intrathecal gabapentin increases interleukin-10 expression and inhibits pro-inflammatory cytokine in a rat model of neuropathic pain.
Intrathecal gabapentin increases interleukin-10 expression and inhibits pro-inflammatory cytokine in a rat model of neuropathic pain.
Journal of Korean medical science (2013-02-13)
Byung-Sang Lee, In-Gu Jun, Sung-Hoon Kim, Jong Yeon Park
ABSTRACT
We examined the possible anti-inflammatory mechanisms of gabapentin in the attenuation of neuropathic pain and the interaction between the anti-allodynic effects of gabapentin and interleukin-10 (IL-10) expression in a rat model of neuropathic pain. The anti-allodynic effect of intrathecal gabapentin was examined over a 7-day period. The anti-allodynic effects of IL-10 was measured, and the effects of anti-IL-10 antibody on the gabapentin were assessed. On day 7, the concentrations of pro-inflammatory cytokines and IL-10 were measured. Gabapentin produced an anti-allodynic effect over the 7-day period, reducing the expression of pro-inflammatory cytokines but increasing the expression of IL-10 (TNF-α, 316.0 ± 69.7 pg/mL vs 88.8 ± 24.4 pg/mL; IL-1β, 1,212.9 ± 104.5 vs 577.4 ± 97.1 pg/mL; IL-6, 254.0 ± 64.8 pg/mL vs 125.5 ± 44.1 pg/mL; IL-10, 532.1 ± 78.7 pg/mL vs 918.9 ± 63.1 pg/mL). The suppressive effect of gabapentin on pro-inflammatory cytokine expression was partially blocked by the anti-IL-10 antibody. Expression of pro-inflammatory cytokines was significantly attenuated by daily injections of IL-10. The anti-allodynic effects of gabapentin may be caused by upregulation of IL-10 expression in the spinal cord, which leads to inhibition of the expression of pro-inflammatory cytokines in the spinal cords.
Sigma-Aldrich
Interleukin-10 from rat, >95% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution