Merck
The therapeutic potential of monoamine oxidase inhibitors.
Nature reviews. Neuroscience (2006-03-23)
Moussa B H Youdim, Dale Edmondson, Keith F Tipton
ABSTRACT
Monoamine oxidase inhibitors were among the first antidepressants to be discovered and have long been used as such. It now seems that many of these agents might have therapeutic value in several common neurodegenerative conditions, independently of their inhibition of monoamine oxidase activity. However, many claims and some counter-claims have been made about the physiological importance of these enzymes and the potential of their inhibitors. We evaluate these arguments in the light of what we know, and still have to learn, of the structure, function and genetics of the monoamine oxidases and the disparate actions of their inhibitors.
Sigma-Aldrich
Monoamine Oxidase B human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
Monoamine Oxidase A human, recombinant, expressed in baculovirus infected BTI insect cells