Merck
  • Altered adrenergic response in myocytes bordering a chronic myocardial infarction underlies in vivo triggered activity and repolarization instability.
Altered adrenergic response in myocytes bordering a chronic myocardial infarction underlies in vivo triggered activity and repolarization instability.
The Journal of physiology (2020-01-05)
Eef Dries, Matthew Amoni, Bert Vandenberk, Daniel M Johnson, Guillaume Gilbert, Chandan K Nagaraju, Rosa Doñate Puertas, Mouna Abdesselem, Demetrio J Santiago, H Llewelyn Roderick, Piet Claus, Rik Willems, Karin R Sipido
ABSTRACT
Ventricular arrhythmias are a major complication after myocardial infarction (MI), associated with sympathetic activation. The structurally heterogeneous peri-infarct zone is a known substrate, but the functional role of the myocytes is less well known. Recordings of monophasic action potentials in vivo reveal that the peri-infarct zone is a source of delayed afterdepolarizations (DADs) and has a high beat-to-beat variability of repolarization (BVR) during adrenergic stimulation (isoproterenol, ISO). Myocytes isolated from the peri-infarct region have more DADs and spontaneous action potentials, with spontaneous Ca2+ release, under ISO. These myocytes also have reduced repolarization reserve and increased BVR. Other properties of post-MI remodelling are present in both peri-infarct and remote myocytes. These data highlight the importance of altered myocyte adrenergic responses in the peri-infarct region as source and substrate of post-MI arrhythmias. Ventricular arrhythmias are a major early complication after myocardial infarction (MI). The heterogeneous peri-infarct zone forms a substrate for re-entry while arrhythmia initiation is often associated with sympathetic activation. We studied the mechanisms triggering these post-MI arrhythmias in vivo and their relation to regional myocyte remodelling. In pigs with chronic MI (6 weeks), in vivo monophasic action potentials were simultaneously recorded in the peri-infarct and remote regions during adrenergic stimulation with isoproterenol (isoprenaline; ISO). Sham animals served as controls. During infusion of ISO in vivo, the incidence of delayed afterdepolarizations (DADs) and beat-to-beat variability of repolarization (BVR) was higher in the peri-infarct than in the remote region. Myocytes isolated from the peri-infarct region, in comparison to myocytes from the remote region, had more DADs, associated with spontaneous Ca2+ release, and a higher incidence of spontaneous action potentials (APs) when exposed to ISO (9.99 ± 4.2 vs. 0.16 ± 0.05 APs/min, p = 0.004); these were suppressed by CaMKII inhibition. Peri-infarct myocytes also had reduced repolarization reserve and increased BVR (26 ± 10 ms vs. 9 ± 7 ms, P < 0.001), correlating with DAD activity. In contrast to these regional distinctions under ISO, alterations in Ca2+ handling at baseline and myocyte hypertrophy were present throughout the left ventricle (LV). Expression of some of the related genes was, however, different between the regions. In conclusion, altered myocyte adrenergic responses in the peri-infarct but not the remote region provide a source of triggered activity in vivo and of repolarization instability amplifying the substrate for re-entry. These findings stimulate further exploration of region-specific therapies targeting myocytes and autonomic modulation.
Product Description
Millipore
ReBlot Plus Strong Antibody Stripping Solution, 10x
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture