
Flavoxate impurity A

Synonym(s):
3-Methyl-4-oxo-2-phenyl-4H-1-benzopyran-8-carboxylic acid
Empirical Formula (Hill Notation):
C17H12O4
CAS Number:
Molecular Weight:
280.27
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing