GC Analysis of Permanent Gases and Light Hydrocarbons on Supel-Q PLOT
![GC Analysis of Permanent Gases and Light Hydrocarbons on Supel-Q PLOT suitable for GC](/deepweb/assets/sigmaaldrich/product/images/767/443/8c092503-1779-4334-a90d-77f740d1caa4/800/8c092503-1779-4334-a90d-77f740d1caa4.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Supel-Q PLOT, 30 m × 0.53 mm I.D. (25462)
oven
35 °C (3 min), 16 °C/min to 250 °C
inj. temp.
200 °C
detector
TCD, 250 °C
carrier gas
helium, 3 mL/min
injection
0.1 μL
sample
9-Component permanent gas a light hydrocarbon standard, each analyte at 1.0% in nitrogen
Description
Legal Information
null