Merck
LC/MS Analysis of Omeprazole and Metabolites on Ascentis® Express HILIC
LC/MS Analysis of Omeprazole and Metabolites on Ascentis® Express HILIC application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express HILIC, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 10 cm × 2.1 mm
standard
Product No.
Description
Pricing
Phenacetin
≥98.0% (HPLC)
Omeprazole sulfone
analytical standard
Omeprazole
analytical standard
CONDITIONS
column
Ascentis Express HILlC, 10 cm x 2.1 mm I.D., 2.7 μm particles (53939-U)
mobile phase
[A] 5 mM ammonium formate in water, pH 6.5; [B] 5 mM ammonium formate in acetonitrile, pH 6.5; (2:98, A:B)
flow rate
0.3 mL/min
column temp.
35 °C
detector
ESI(+), m/z 50-400
injection
0.1 μL
sample
1 mg/L each in mobile phase
Description
Analysis Note
This application demonstrates the suitability of the Ascentis Express HILIC for the analysis of omeprazole metabolites.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany