Merck
US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
US EPA Method 8270 (PAH only): GC Analysis of PAHs on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 20 m × 0.18 mm, df 0.18 μm
CONDITIONS
column
SLB-5ms, 20 m x 0.18 mm I.D., 0.18 μm (28564-U)
oven
50 °C, 25 °C/min to 250 °C, 10 °C/min to 330 °C
inj. temp.
300 °C
MSD interface
300 °C
scan range
m/z 45 - 450
carrier gas
helium, 1.5 mL/min, constant
injection
helium, 1.5 mL/min, constant
liner
2 mm I.D. FocusLiner
sample
Each analyte at 5 μg/mL in methylene chloride
Description
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany