GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SLB®-IL100
![GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SLB®-IL100 suitable for GC](/deepweb/assets/sigmaaldrich/product/images/177/593/9460e75a-76d3-4b50-839d-c16115f11a77/800/9460e75a-76d3-4b50-839d-c16115f11a77.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL100, 60 m x 0.25 mm I.D., 0.20 μm (28886-U)
oven
170 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 cm/sec
injection
1 μL 50:1 split
liner
4 mm I.D., split, cup design
sample
Mixture of C18:1, C18:2, and C18:3 FAMEs in methylene chloride
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany