GC Analysis of Blood Alcohols on a Packed Column (5% Carbowax® 20M on 60/80 Carbopack™ B)
![GC Analysis of Blood Alcohols on a Packed Column (5% Carbowax® 20M on 60/80 Carbopack™ B) suitable for GC](/deepweb/assets/sigmaaldrich/product/images/265/708/2eb8d955-e469-449a-8770-67a685735de6/800/2eb8d955-e469-449a-8770-67a685735de6.jpg)
Materials
standard
Product No.
Description
Pricing
CONDITIONS
column
6 ft.x 2 mm I.D. glass
packing
5% Carbowax 20M on 60/80 Carbopack B
oven
85 °C
detector
FID
carrier gas
helium, 20 mL/min
injection
1 μL
sample
0.05-0.10% each analyte in water
Description
Legal Information
Carbopack is a trademark of Merck KGaA, Darmstadt, Germany
Carbowax is a registered trademark of Union Carbide