Merck
  • SERS-fluorescence bimodal nanoprobes for in vitro imaging of fatty acid responsive receptor GPR120.
SERS-fluorescence bimodal nanoprobes for in vitro imaging of fatty acid responsive receptor GPR120.
Analytical methods : advancing methods and applications (2018-02-17)
Lifu Xiao, Abdul K Parchur, Timothy A Gilbertson, Anhong Zhou
ABSTRACT
G-protein-coupled receptor 120 (GPR120), as a member of the rhodopsin family of G-protein-coupled receptors, has been shown to function as a sensor for dietary fat in the gustatory and digestive systems. Its specific role in the chemoreception of fatty acids, which is thought to be crucial in understanding the mechanism surrounding the control of fat intake and, accordingly, in the treatment of obesity, remains unclear. Here we report a novel surface-enhanced Raman spectroscopy (SERS)-fluorescence bimodal microscopic technique for detection and imaging of GPR120 in single living cells. CaMoO
Sigma-Aldrich
Europium(III) nitrate hydrate, 99.99% trace metals basis