Merck
  • Phenoxodiol Increases Cisplatin Sensitivity in Ovarian Clear Cancer Cells Through XIAP Down-regulation and Autophagy Inhibition.
Phenoxodiol Increases Cisplatin Sensitivity in Ovarian Clear Cancer Cells Through XIAP Down-regulation and Autophagy Inhibition.
Anticancer research (2017-12-27)
Morikazu Miyamoto, Masashi Takano, Tadashi Aoyama, Hiroaki Soyama, Hiroki Ishibashi, Kento Kato, Hideki Iwahashi, Kazuki Takasaki, Mika Kuwahara, Hiroko Matuura, Takahiro Sakamoto, Tomoyuki Yoshikawa, Kenichi Furuya
ABSTRACT
To investigate whether XIAP down-regulation and autophagy inhibition sensitize ovarian clear cell cancer cells to cisplatin. The ovarian clear cancer cell line KK was used for in vitro analysis. Hydroxychloroquine (HCQ) and phenoxodiol (PXD) or embelin were used as autophagy and XIAP inhibitors, respectively. Non-specific and XIAP-specific siRNAs were transfected using Lipofectamine. Cytotoxicity was assessed by MTT assays. Protein expression was confirmed by western blotting. In KK, down-regulation of XIAP using specific siRNAs together with HCQ treatment enhanced the anti-tumor effect of cisplatin. Although embelin sensitized KK to cisplatin through XIAP down-regulation, it induced autophagy. However, PXD increased cisplatin sensitivity through XIAP down-regulation and autophagy inhibition. Expression of Atg7, Atg12, and Beclin 1 was decreased after PXD treatment. PXD increased cisplatin sensitivity through XIAP down-regulation and autophagy inhibition and could be a new candidate for ovarian clear cell carcinoma treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human XIAP
Pricing and availability is not currently available.