Merck
  • Antimalarial activity of molecules interfering with Plasmodium falciparum phospholipid metabolism. Structure-activity relationship analysis.
Antimalarial activity of molecules interfering with Plasmodium falciparum phospholipid metabolism. Structure-activity relationship analysis.
Journal of medicinal chemistry (1997-11-14)
M Calas, G Cordina, J Bompart, M Ben Bari, T Jei, M L Ancelin, H Vial
ABSTRACT
A series of 80 compounds, primary, secondary, and tertiary amines and quaternary ammonium and bisammonium salts, most of them synthesized as potential choline or ethanolamine analogs, were tested against the in vitro growth of Plasmodium falciparum, the human malaria parasite. They were active over the 10(-3)-10(-8) M concentration range. A structure-activity relationship study was carried out using autocorrelation vectors as structural descriptors, and multidimensional analysis. Principal component analysis, ascending hierarchical classification, and stepwise discriminant analysis showed that both the size and shape of the molecule were essential for antimalarial potency, making the lipophilicity and electronegativity distribution in the molecular space essential. Using the autocorrelogram describing the molecular shape and the electronegativity distribution on the molecular graph, 98% of the molecules were correctly classified either as poorly active or active with only three explanatory variables. The most active compounds were quaternary ammoniums salts whose nitrogen atom had only one long lipophilic chain of 11 or 12 methylene groups (E5, E6, E10, E13, E20, E21, E22, E23, F4, F8), or the bisammoniums whose polar heads were linked by linear alkyl chains of 10 to 12 carbon atoms (G4, G23). The hydroxyethyl group of choline was not very beneficial, whereas the charge and substitutions of nitrogen (aimed at increasing lipophilicity) were essential for optimal interactions. A crude topographic model of the ligand (choline) binding site was thus drawn up.
Sigma-Aldrich
Trimethyloctadecylammonium chloride, ≥95.0% (calc. on dry substance, T)