Merck
  • Endogenous, regulatory cysteine sulfenylation of ERK kinases in response to proliferative signals.
Endogenous, regulatory cysteine sulfenylation of ERK kinases in response to proliferative signals.
Free radical biology & medicine (2017-08-28)
Jeremiah D Keyes, Derek Parsonage, Rama D Yammani, LeAnn C Rogers, Chelsea Kesty, Cristina M Furdui, Kimberly J Nelson, Leslie B Poole
ABSTRACT
ERK-dependent signaling is key to many pathways through which extracellular signals are transduced into cell-fate decisions. One conundrum is the way in which disparate signals induce specific responses through a common, ERK-dependent kinase cascade. While studies have revealed intricate ways of controlling ERK signaling through spatiotemporal localization and phosphorylation dynamics, additional modes of ERK regulation undoubtedly remain to be discovered. We hypothesized that fine-tuning of ERK signaling could occur by cysteine oxidation. We report that ERK is actively and directly oxidized by signal-generated H
Sigma-Aldrich
Catalase−polyethylene glycol, lyophilized powder, ~40,000 units/mg protein