Merck
  • Molecular conductors from bis(ethylenedithio)tetrathiafulvalene with tris(oxalato)rhodate.
Molecular conductors from bis(ethylenedithio)tetrathiafulvalene with tris(oxalato)rhodate.
Dalton transactions (Cambridge, England : 2003) (2017-07-14)
Lee Martin, Alexander L Morritt, Jordan R Lopez, Yasuhiro Nakazawa, Hiroki Akutsu, Shusaku Imajo, Yoshihiko Ihara, Bin Zhang, Yan Zhang, Yanjun Guo
ABSTRACT
This article reports a family of new radical-cation salts of bis(ethylenedithio)tetrathiafulvalene (BEDT-TTF) with tris(oxalato)rhodate: three salts with the formula β''-(BEDT-TTF)
Sigma-Aldrich
Bis(ethylenedithio)tetrathiafulvalene, 98%
Pricing and availability is not currently available.