Merck
  • Application of metabolic controls for the maximization of lipid production in semicontinuous fermentation.
Application of metabolic controls for the maximization of lipid production in semicontinuous fermentation.
Proceedings of the National Academy of Sciences of the United States of America (2017-06-21)
Jingyang Xu, Nian Liu, Kangjian Qiao, Sebastian Vogg, Gregory Stephanopoulos
ABSTRACT
Acetic acid can be generated through syngas fermentation, lignocellulosic biomass degradation, and organic waste anaerobic digestion. Microbial conversion of acetate into triacylglycerols for biofuel production has many advantages, including low-cost or even negative-cost feedstock and environmental benefits. The main issue stems from the dilute nature of acetate produced in such systems, which is costly to be processed on an industrial scale. To tackle this problem, we established an efficient bioprocess for converting dilute acetate into lipids, using the oleaginous yeast
Sigma-Aldrich
Glyceryl triheptadecanoate, ≥99%