Merck
  • BMS-536924 reverses IGF-IR-induced transformation of mammary epithelial cells and causes growth inhibition and polarization of MCF7 cells.
BMS-536924 reverses IGF-IR-induced transformation of mammary epithelial cells and causes growth inhibition and polarization of MCF7 cells.
Clinical cancer research : an official journal of the American Association for Cancer Research (2009-01-02)
Beate C Litzenburger, Hyun-Jung Kim, Isere Kuiatse, Joan M Carboni, Ricardo M Attar, Marco M Gottardis, Craig R Fairchild, Adrian V Lee
ABSTRACT
This study aimed to test the ability of a new insulin-like growth factor receptor (IGF-IR) tyrosine kinase inhibitor, BMS-536924, to reverse the ability of constitutively active IGF-IR (CD8-IGF-IR) to transform MCF10A cells, and to examine the effect of the inhibitor on a range of human breast cancer cell lines. CD8-IGF-IR-MCF10A cells were grown in monolayer culture, three-dimensional (3D) culture, and as xenografts, and treated with BMS-536924. Proliferation, cell cycle, polarity, and apoptosis were measured. Twenty-three human breast cancer cell lines were treated in monolayer culture with BMS-536924, and cell viability was measured. MCF7, MDA-MB-231, and MDA-MB-435 were treated with BMS-536924 in monolayer and 3D culture, and proliferation, migration, polarity, and apoptosis were measured. Treatment of CD8-IGF-IR-MCF10A cells grown in 3D culture with BMS-536924 caused a blockade of proliferation, restoration of apical-basal polarity, and enhanced apoptosis, resulting in a partial phenotypic reversion to normal acini. In monolayer culture, BMS-536924 induced a dose-dependent inhibition of proliferation, with an accumulation of cells in G(0)/G(1,), and completely blocked CD8-IGF-IR-induced migration, invasion, and anchorage-independent growth. CD8-IGF-IR-MCF10A xenografts treated with BMS-536924 (100 mg/kg/day) showed a 76% reduction in xenograft volume. In a series of 23 human breast cancer cell lines, BMS-536924 inhibited monolayer proliferation of 16 cell lines. Most strikingly, treatment of MCF7 cells grown in 3D culture with BMS-536924 caused blockade of proliferation, and resulted in the formation of hollow polarized lumen. These results show that the new small molecule BMS-536924 is an effective inhibitor of IGF-IR, causing a reversion of an IGF-IR - mediated transformed phenotype.