Merck
  • A traceable reference for direct comparative assessment of total naphthenic acid concentrations in commercial and acid extractable organic mixtures derived from oil sands process water.
A traceable reference for direct comparative assessment of total naphthenic acid concentrations in commercial and acid extractable organic mixtures derived from oil sands process water.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2016-12-03)
Pamela Brunswick, L Mark Hewitt, Richard A Frank, Marcus Kim, Graham van Aggelen, Dayue Shang
ABSTRACT
The advantage of using naphthenic acid (NA) mixtures for the determination of total NA lies in their chemical characteristics and identification of retention times distinct from isobaric interferences. However, the differing homolog profiles and unknown chemical structures of NA mixtures do not allow them to be considered a traceable reference material. The current study provides a new tool for the comparative assessment of different NA mixtures by direct reference to a single, well-defined and traceable compound, decanoic-d
Sigma-Aldrich
Naphthenic acid, technical
Sigma-Aldrich
Decanoic-d19 acid, 98 atom % D, 98% (CP)