Merck
  • Adenovirus-induced extracellular signal-regulated kinase phosphorylation during the late phase of infection enhances viral protein levels and virus progeny.
Adenovirus-induced extracellular signal-regulated kinase phosphorylation during the late phase of infection enhances viral protein levels and virus progeny.
Cancer research (2006-02-03)
Michael Schümann, Matthias Dobbelstein
ABSTRACT
The Raf/mitogen-activated protein/extracellular signal-regulated kinase (ERK) kinase (MEK)/ERK signaling cascade enhances tumor cell proliferation in many cases. Here, we show that adenovirus type 5, a small DNA tumor virus used in experimental cancer therapy, strongly induces ERK phosphorylation during the late phase of infection. Pharmacologic inhibition of ERK phosphorylation reduced virus recovery by >100-fold. Blocking MEK/ERK signaling affected virus DNA replication and mRNA levels only weakly but strongly reduced the amount of viral proteins, independently of the kinases MNK1 and PKR. Hence, adenovirus induces the oncogenic Raf/MEK/ERK signaling pathway to enhance viral progeny by sustaining the levels of viral proteins. Concerning therapy, our results suggest that the use of Raf/MEK/ERK inhibitors will interfere with the propagation of oncolytic adenoviruses.
Sigma-Aldrich
CGP 57380, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
U0126, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC₅₀ = 72 nM) and MEK2 (IC₅₀ = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Pricing and availability is not currently available.