Merck
  • Glutathione transferases immobilized on nanoporous alumina: flow system kinetics, screening, and stability.
Glutathione transferases immobilized on nanoporous alumina: flow system kinetics, screening, and stability.
Analytical biochemistry (2013-10-26)
Marcus Kjellander, Aslam M A Mazari, Mats Boman, Bengt Mannervik, Gunnar Johansson
ABSTRACT
The previously uncharacterized Drosophila melanogaster Epsilon-class glutathione transferases E6 and E7 were immobilized on nanoporous alumina. The nanoporous anodized alumina membranes were derivatized with 3-aminopropyl-triethoxysilane, and the amino groups were activated with carbonyldiimidazole to allow coupling of the enzymes via ε-amino groups. Kinetic analyses of the immobilized enzymes were carried out in a circulating flow system using CDNB (1-chloro-2,4-dinitrobenzene) as substrate, followed by specificity screening with alternative substrates. A good correlation was observed between the substrate screening data for immobilized enzyme and corresponding data for the enzyme in solution. A limited kinetic study was also carried out on immobilized human GST S1-1 (also known as hematopoietic prostaglandin D synthase). The stability of the immobilized enzymes was virtually identical to that of enzymes in solution, and no leakage of enzyme from the matrix could be observed.