Merck
Minimal change multiple system atrophy: an aggressive variant?
Movement disorders : official journal of the Movement Disorder Society (2015-04-10)
Helen Ling, Yasmine T Asi, Igor N Petrovic, Zeshan Ahmed, L K Prashanth, Lili-Naz Hazrati, Masatoyo Nishizawa, Tetsutaro Ozawa, Anthony Lang, Andrew J Lees, Tamas Revesz, Janice L Holton
ABSTRACT
Glial cytoplasmic inclusions containing α-synuclein are the pathological hallmark of multiple system atrophy (MSA). Minimal change (MC-MSA) is an unusual MSA subtype with neuronal loss largely restricted to the substantia nigra and locus coeruleus. Immunohistochemistry on selected brain regions and semiquantitative assessment were performed on six MC-MSA and eight MSA control cases. More neuronal cytoplasmic inclusions were seen in the caudate and substantia nigra in MC-MSA than in MSA controls (P = 0.002), without any statistical difference in glial cytoplasmic inclusion load in any region. Severe glial cytoplasmic inclusion load was found in the ventrolateral medulla (P = 1.0) and nucleus raphe obscurus (P = 0.4) in both groups. When compared with MSA controls, the three MC-MSA cases who had died of sudden unexpected death had an earlier age of onset (mean: 38 vs. 57.6 y, P = 0.02), a numerically shorter disease duration (mean: 5.3 vs. 8 y, P = 0.2) and a more rapid clinical progression with most of the clinical milestones reached within 3 y of presentation, suggesting an aggressive variant of MSA. Another three MC-MSA cases, who had died of unrelated concurrent diseases, had an age of onset (mean: 57.7 y) and temporal course similar to controls, had less severe neuronal loss and gliosis in the medial and dorsolateral substantia nigra subregions (P < 0.05) than in MSA controls, and could be considered as a unique group with interrupted pathological progression. Significant respiratory dysfunction and early orthostatic hypotension were observed in all MC-MSA cases. Our findings could suggest that α-synuclein-associated oligodendroglial pathology may lead to neuronal dysfunction sufficient to cause clinical symptoms before overt neuronal loss in MSA. © 2015 International Parkinson and Movement Disorder Society.
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Tyrosine, 99%