Merck
  • Assessing the differential action on cancer cells of LDH-A inhibitors based on the N-hydroxyindole-2-carboxylate (NHI) and malonic (Mal) scaffolds.
Assessing the differential action on cancer cells of LDH-A inhibitors based on the N-hydroxyindole-2-carboxylate (NHI) and malonic (Mal) scaffolds.
Organic & biomolecular chemistry (2013-08-30)
Carlotta Granchi, Emilia C Calvaresi, Tiziano Tuccinardi, Ilaria Paterni, Marco Macchia, Adriano Martinelli, Paul J Hergenrother, Filippo Minutolo
ABSTRACT
A head-to-head study of representative examples of N-hydroxyindole-2-carboxylates (NHI) and malonic derivatives (Mal) as LDH-A inhibitors was conducted, comparing the enzyme inhibition potency, cellular uptake, reduction of lactate production in cancer cells and anti-proliferative activity. Among the compounds tested, methyl 1-hydroxy-6-phenyl-4-(trifluoromethyl)-1H-indole-2-carboxylate (2, NHI-2), a methyl ester belonging to the NHI class, displayed optimal properties in the cell-based assays, proving to be an efficient anti-glycolytic agent against cancer cells.
Sigma-Aldrich
NHI-2, ≥98% (HPLC)
Pricing and availability is not currently available.