Merck
  • Depletion of IK causes mitotic arrest through aberrant regulation of mitotic kinases and phosphatases.
Depletion of IK causes mitotic arrest through aberrant regulation of mitotic kinases and phosphatases.
FEBS letters (2014-07-06)
Sunyi Lee, Sora Han, Ae Lee Jeong, Jeong Su Park, Young Yang
ABSTRACT
IK is known to inhibit the expression of major histocompatibility complex (MHC) class II antigen, but other cellular functions of IK remain to be uncovered. In this study, IK depletion caused misalignment of chromosomes through an increase in Aurora A and PLK1 phosphorylation, which was mediated by a decrease in PP1 and PP2A activities. On the other hand, the treatment of a dual inhibitor against CDK and Aurora kinases overrode IK depletion-induced mitotic arrest through the activation of phosphatase activity. These findings imply that IK is an essential protein for achieving correct mitotic progress through the regulation of mitotic kinases and phosphatases.
Sigma-Aldrich
Aurora Kinase A active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)