Merck
New cholinesterase inhibitors from Garcinia atroviridis.
Fitoterapia (2014-06-14)
Wen-Nee Tan, Melati Khairuddean, Keng-Chong Wong, Kooi-Yeong Khaw, Murugaiyah Vikneswaran
ABSTRACT
A triflavanone, Garcineflavanone A (1) and a biflavonol, Garcineflavonol A (2) have been isolated from the stem bark of Garcinia atroviridis (Clusiaceae), collected in Peninsular Malaysia. Their structures were established using one and two-dimensional NMR, UV, IR and mass spectrometry and evaluated in vitro for their acetylcholinesterase (AChE) and butyrylcholinesterase (BChE) enzymes inhibitory activity. Molecular docking studies of the isolated compounds were performed using docking procedure of AutoDock to disclose the binding interaction and orientation of these molecules into the active site gorge.
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups