Merck
  • Synthesis and anti-HIV and anti-HBV activities of 2'-fluoro-2', 3'-unsaturated L-nucleosides.
Synthesis and anti-HIV and anti-HBV activities of 2'-fluoro-2', 3'-unsaturated L-nucleosides.
Journal of medicinal chemistry (1999-04-10)
K Lee, Y Choi, E Gullen, S Schlueter-Wirtz, R F Schinazi, Y C Cheng, C K Chu
ABSTRACT
The synthesis of L-nucleoside analogues containing 2'-vinylic fluoride was accomplished by direct condensation method, and their anti-HIV and anti-HBV activities were evaluated in vitro. The key intermediate 8, the sugar moiety of our target compounds, was prepared from 1,2-O-isopropylidene-L-glyceraldehyde via (R)-2-fluorobutenolide intermediate 5 in five steps. Coupling of the acetate 8 with the appropriate heterocycles (silylated uracil, thymine, N4-benzoylcytosine, N4-benzoyl-5-fluorocytosine, 6-chloropurine, and 6-chloro-2-fluoropurine) in the presence of Lewis acid afforded a series of 2'-fluorinated L-nucleoside analogues (15-18, 23-26, 36-45). The newly synthesized compounds were evaluated for their antiviral activities against HIV-1 in human peripheral blood mononuclear (PBM) cells and HBV in 2.2.15 cells. Cytosine 23, 5-fluorocytosine 25, and adenine 36 derivatives exhibited moderate to potent anti-HIV (EC50 0.51, 0.17, and 1.5 microM, respectively) and anti-HBV (EC50 0.18, 0.225, and 1.7 microM, respectively) activities without significant cytotoxicity up to 100 microM in human PBM, Vero, CEM, and HepG2 cells.
Sigma-Aldrich
N4-Benzoylcytosine, 98%