Merck
GRASP-1 is a neuronal scaffold protein for the JNK signaling pathway.
FEBS letters (2007-09-01)
Bing Ye, Wei-ping Yu, Gareth M Thomas, Richard L Huganir
ABSTRACT
GRASP-1 is a neuronally enriched protein that interacts with the AMPA-type glutamate receptor/GRIP complex. GRASP-1 can be cleaved by Caspase-3 in both normal and ischemic brains although the functional significance of this cleavage remains elusive. We investigated signal transduction pathways that might lie downstream of GRASP-1 and found that GRASP-1 potently activates JNK pathway signaling, with no effect on ERK signaling. Such JNK pathway activating activity requires binding of GRASP-1 to both JNK and the upstream JNK pathway activator MEKK-1. Furthermore, mutations that prevent Caspase 3-cleavage of GRASP-1 dramatically inhibit the JNK pathway activating activity of GRASP-1, suggesting a novel link between Caspase-3 activation and JNK pathway signaling. These results suggest that GRASP-1 serves as a scaffold protein to facilitate MEKK-1 activation of JNK signaling in neurons.