Merck
  • Synthesis and pharmacology of metabolically stable tert-butyl ethers of morphine and levorphanol.
Synthesis and pharmacology of metabolically stable tert-butyl ethers of morphine and levorphanol.
Journal of medicinal chemistry (1982-10-01)
E Mohacsi, W Leimgruber, H Baruth
ABSTRACT
3-O-tert-Butylmorphine (5) was prepared from 6-O-acetylmorphine (3) via alkylation with N,N-dimethylformamide di-tert-butyl acetal, followed by hydrolytic removal of the 3-(dimethylamino)-2-propenoate group. The same process was used to prepare the tert-butyl ether of levorphanol (6), (-)-3-tert-butoxy-N-methylmorphinan (8). Both 5 and 8 exhibited in vitro affinity for the opiate receptor comparable to codeine and had analgesic properties in the writhing test. Only 5 exhibited activity in the tail-flick procedure and neither compound showed significant antitussive activity.
Sigma-Aldrich
N,N-Dimethylformamide di-tert-butyl acetal, technical, ≥90% (GC)
Pricing and availability is not currently available.