Merck
  • Synthesis and evaluation of tricyclic derivatives containing a non-aromatic amide as inhibitors of poly(ADP-ribose)polymerase-1 (PARP-1).
Synthesis and evaluation of tricyclic derivatives containing a non-aromatic amide as inhibitors of poly(ADP-ribose)polymerase-1 (PARP-1).
Bioorganic & medicinal chemistry letters (2010-03-02)
Chun-Ho Park, Kwangwoo Chun, Bo-Young Joe, Ji-Seon Park, Young-Chul Kim, Ji-Soo Choi, Dong-Kyu Ryu, Seong-Ho Koh, Goang Won Cho, Seung Hyun Kim, Myung-Hwa Kim
ABSTRACT
Highly potent poly(ADP-ribose)polymerase-1 (PARP-1) inhibitors, including 9-hydroxy-1,2-dihydro-4H-thiopyrano[3,4-c]quinolin-5(6H)-one derivatives with a non-aromatic A-ring, were synthesized. Among the derivatives, 12a showed low nanomolar enzyme and cellular activity (IC(50) = 42 nM, ED(50) = 220 nM) with good water solubility. Further, 12a exhibited microsomal stability in vitro and brain permeability in vivo.
Sigma-Aldrich
Ethyl 4-isocyanatobenzoate, 97%