MilliporeSigma
  • Leonurine (SCM-198) improves cardiac recovery in rat during chronic infarction.
Leonurine (SCM-198) improves cardiac recovery in rat during chronic infarction.
European journal of pharmacology (2010-09-22)
XinHua Liu, LiLong Pan, QiHai Gong, YiZhun Zhu
ABSTRACT
Leonurine, an alkaloid typically found in Herba leonuri, is known to have both antioxidant and cardioprotective properties. In the present study, we investigated the cardioprotective mechanism of leonurine the in vivo rat model of chronic myocardial ischemia and in vitro H9c2 cardiac myocyte model of oxidative stress. Myocardial ischemia was induced by ligating the left anterior descending coronary artery. Rats were divided into sham, myocardial ischemia+saline, and myocardial ischemia+leonurine (15 mg/kg/day). Cardiac function was recorded by catheterization. Apoptosis-related factor vascular endothelial growth factor (VEGF), survivin, Bcl-2 and Bax and pro-survival signaling pathways Akt, hypoxia inducible factor (HIF)-1α were measured by Western blotting or RT-PCR. Our results showed leonurine significantly improved myocardial function as evidenced by the decreased left ventricle end-diastolic pressure and the increased +dP/dt. Interestingly, leonurine increased the phosphorylation of Akt, the protein and gene expression of Bcl-2, but it reduced the protein and gene expression of Bax in vivo. Meanwhile leonurine significantly increased Akt phosphorylation in a concentration-dependent manner in H9c2 cardiac myocyte induced by oxidative stress in vitro, which was abolished by a phosphoinositide 3-kinase (PI3K) inhibitor, LY294002. Furthermore, leonurine not only increased the expression of HIF-1α but also the expression of survivin and VEGF. The results of present study demonstrated, for the first time that leonurine has potent anti-apoptotic effects after chronic myocardial ischemia mediated by activating the PI3K/Akt signaling pathway. Angiogenic mechanisms may be partially responsible for such an effect, which needs to be studied further.
Sigma-Aldrich
Leonurine, ≥98% (HPLC)