Merck
  • Preparation, physicochemical characterization and pharmacological study of novel ruthenium(III) complexes with imidazole and benzimidazole derivatives.
Preparation, physicochemical characterization and pharmacological study of novel ruthenium(III) complexes with imidazole and benzimidazole derivatives.
Arzneimittel-Forschung (2001-10-20)
A Nikolova, D Ivanov, R Buyukliev, S Konstantinov, M Karaivanova
ABSTRACT
Complex compounds of ruthenium(III) with 1,2-dimethylimidazole (CAS 1739-84-0), 2-phenylimidazole (CAS 670-96-2) and 2-aminobenzimidazole (CAS 934-32-7) were prepared and were characterised by physicochemical methods. Coordination sites were determined. The complexes were tested for cytotoxic activity using MTT (3-(4,5-dimethyithiazol-2-yl)-2,5-diphenyltetrazolium bromide) dye-reduction assay and the values LD50 were evaluated.
Sigma-Aldrich
2-Phenylimidazole, 98%