Merck
  • Experimental (NMR) and Computational (MD) Studies on the Inclusion Complexes of 1-Bromoadamantane with alpha-, beta-, and gamma-Cyclodextrin.
Experimental (NMR) and Computational (MD) Studies on the Inclusion Complexes of 1-Bromoadamantane with alpha-, beta-, and gamma-Cyclodextrin.
The Journal of organic chemistry (1996-10-04)
P. M. Ivanov, D. Salvatierra, C. Jaime
ABSTRACT
The inclusion complexes between the most commonly used cyclodextrins (alpha-, beta-, and gamma-CD) and 1-bromoadamantane were prepared and studied experimentally by NMR methods and by molecular dynamics simulations (AMBER force field) with solvation. The NMR results suggest host/guest ratios of 2:1, 1:1, and 1:1 for the complexes with alpha-, beta-, and gamma-cyclodextrin, respectively, as well as defined geometries for the complexes. Averaged geometrical data from the molecular dynamics simulations agree with the complexation geometries deduced experimentally.
Sigma-Aldrich
1-Bromoadamantane, 99%