- Studies on a child suspected of having a dficiency in 3-hydroxy-3-methylglutaryl-Co A lyase.
Studies on a child suspected of having a dficiency in 3-hydroxy-3-methylglutaryl-Co A lyase.
The urine of a child who presented with hyperammonemia was found to contain elevated levels of 3-hydroxy-3-methylglutaric acid, 3-methylglutaconic acid and 3-methylglutaric acid. An increased excretion of these organic acids has been reported previously in a child with 3-hydroxy-3-methylglutaryl-CoA lyase deficiency. Enzyme studies using cultured fibroblasts from this patient, however, indicated that the 3-hydroxy-3-methylglutaryl-CoA lyase activity was not markedly reduced.