- [Exacerbation of an affective psychosis after terminating a decade of benzodiazepine treatment. Which therapeutic procedure is sensible?].
[Exacerbation of an affective psychosis after terminating a decade of benzodiazepine treatment. Which therapeutic procedure is sensible?].
Der Nervenarzt (1994-09-01)
M Soyka, R Lehle, H Hippius
PMID7991010
ABSTRACT
We report the case of a 61-year old patient with an affective disorder who had been treated with benzodiazepines in low dosages over a 16 year period. This treatment had been prescribed by his physician following several depressive episodes. During this time, the patient remained able to work and exhibited little psychopathological symptomatology. Following discontinuation of medication, however, the depressive phases resumed and were particularly intractable. The clinical implications of this case and the therapeutic strategies for approaching patients with long-term benzodiazepine treatment are discussed.
Product Description
Supelco
Prazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, CerilliantĀ®