Merck
A mutant HpaII methyltransferase functions as a mutator enzyme.
Nucleic acids research (1995-11-11)
J C Shen, J M Zingg, A S Yang, C Schmutte, P A Jones
ABSTRACT
DNA (cytosine-5)-methyltransferases can cause deamination of cytosine when the cofactor S-adenosylmethionine (AdoMet) is limiting and thus function as sequence-specific C-->U mutator enzymes. Here we explored whether mutations causing inactivation of the cofactor binding activity of the HpaII methyltransferase, thus mimicking conditions of limiting AdoMet concentration, could convert a DNA methyltransferase to a C-->U mutator enzyme. We created two mutator enzymes from the HpaII methyltransferase (F38S and G40D) which both showed enhanced cytosine deamination activities in vitro and in vivo. Interestingly, the G:U mispairs generated by these enzymes were not repaired completely in bacteria equipped with uracil-DNA glycosylase-initiated repair machinery, giving rise to a potent mutator phenotype. This is the first report showing the creation of mutator enzymes from a DNA methyltransferase and the demonstration of their mutagenicity in living cells.
Sigma-Aldrich
5−Fluoro−2′−deoxycytidine, ≥98% (HPLC), powder