Merck
Alkyllead compounds and their environmental toxicology.
Metal ions in life sciences (2010-09-30)
Henry G Abadin, Hana R Pohl
ABSTRACT
Alkyllead compounds are man-made compounds in which a carbon atom of one or more organic molecules is bound to a lead atom. Tetraethyllead and tetramethyllead are the most common alkyllead compounds that were used primarily as gasoline additives for many years. Consequently, auto emissions have accounted for a major part of lead environmental pollution. Alkyllead compounds can readily enter living organisms as they are well absorbed via all major routes of entry. Because of their lipid solubility, the alkylleads can also readily cross the blood-brain barrier. The toxicokinetic information on organic lead can be used as biomarkers of exposure for monitoring exposed individuals. The organic alkyllead compounds are more toxic than the inorganic forms of lead. Neurotoxicity is the predominant effect of lead (both for organic and inorganic forms), although lead affects almost every organ of the body. The use of alkyllead compounds has declined over the last 20 years, due to the worldwide effort to eliminate the use of leaded gasoline. This achievement can be viewed as a great accomplishment of public health preventive measures.