Merck
  • Expedient synthesis of cyclopropane alpha-amino acids by the catalytic asymmetric cyclopropanation of alkenes using iodonium ylides derived from methyl nitroacetate.
Expedient synthesis of cyclopropane alpha-amino acids by the catalytic asymmetric cyclopropanation of alkenes using iodonium ylides derived from methyl nitroacetate.
Journal of the American Chemical Society (2005-12-22)
Benoît Moreau, André B Charette
ABSTRACT
A highly enantioselective (up to 97.5% ee) and diastereoselective (95:5 dr trans/cis) Cu(I)-catalyzed cyclopropanation of alkenes using phenyliodonium ylide generated in situ from iodosobenzene and methyl nitroacetate is reported. The cyclopropanation took place with high enantioselectivity for a wide range of alkenes, and the reaction was performed at room temperature. 1-Nitrocyclopropyl esters are versatile building blocks to access the corresponding cyclopropane amino esters and aminocyclopropanes in two and three steps, respectively, from commercially available products.
Sigma-Aldrich
Methyl nitroacetate, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
(+)-2,2′-Isopropylidenebis[(4R)-4-phenyl-2-oxazoline], 97%
Pricing and availability is not currently available.