Merck
  • Spectroscopic and kinetic studies of interactions of calf spleen purine nucleoside phosphorylase with 8-azaguanine, and its 9-(2-phosphonylmethoxyethyl) derivative.
Spectroscopic and kinetic studies of interactions of calf spleen purine nucleoside phosphorylase with 8-azaguanine, and its 9-(2-phosphonylmethoxyethyl) derivative.
Nucleosides, nucleotides & nucleic acids (2005-10-27)
Jacek Wierzchowski, Katarzyna Stepniak, Agnieszka Bzowska, David Shugar
ABSTRACT
Spectroscopic and kinetic studies of interactions of calf spleen purine nucleoside phosphorylase with 8-azaguanine, an excellent fluorescent/fluorogenic substrate for the synthetic pathway of the reaction, and its 9-(2-phosphonylmethoxyethyl) derivative, a bisubstrate analogue inhibitor, were carried out. The goal was to clarify the catalytic mechanism of the enzymatic reaction by identification of ionic/tautomeric forms of these ligands in the complex with PNP.