Merck
  • PPARγ regulates expression of carbohydrate sulfotransferase 11 (CHST11/C4ST1), a regulator of LPL cell surface binding.
PPARγ regulates expression of carbohydrate sulfotransferase 11 (CHST11/C4ST1), a regulator of LPL cell surface binding.
PloS one (2013-05-23)
Ismayil Tasdelen, Ruud Berger, Eric Kalkhoven
ABSTRACT
The transcription factor PPARγ is the key regulator of adipocyte differentiation, function and maintenance, and the cellular target of the insulin-sensitizing thiazolidinediones. Identification and functional characterization of genes regulated by PPARγ will therefore lead to a better understanding of adipocyte biology and may also contribute to the development of new anti-diabetic drugs. Here, we report carbohydrate sulfotransferase 11 (Chst11/C4st1) as a novel PPARγ target gene. Chst11 can sulphate chondroitin, a major glycosaminoglycan involved in development and disease. The Chst11 gene contains two functional intronic PPARγ binding sites, and is up-regulated at the mRNA and protein level during 3T3-L1 adipogenesis. Chst11 knockdown reduced intracellular lipid accumulation in mature adipocytes, which is due to a lowered activity of lipoprotein lipase, which may associate with the adipocyte cell surface through Chst11-mediated sulfation of chondroitin, rather than impaired adipogenesis. Besides directly inducing Lpl expression, PPARγ may therefore control lipid accumulation by elevating the levels of Chst11-mediated proteoglycan sulfation and thereby increasing the binding capacity for Lpl on the adipocyte cell surface.
Sigma-Aldrich
LPL Activity Assay Kit, Supplied by Roar Biomedical, Inc.
Pricing and availability is not currently available.