Merck
  • Synthesis and evaluation of antiviral activity of L-acosamine and L-ristosamine nucleosides of furanose configuration.
Synthesis and evaluation of antiviral activity of L-acosamine and L-ristosamine nucleosides of furanose configuration.
Acta chemica Scandinavica (Copenhagen, Denmark : 1989) (1991-07-01)
J Lau, E B Pedersen, C M Nielsen
ABSTRACT
Mercuric-catalyzed hydrolysis of acetylated L-rhamnal 1 gives an alpha,beta-unsaturated aldehyde 2. 1,4-Addition of DBU-phthalimide salt with concomitant acetyl shift resulted in L-ribo and L-arabino isomers of 5-O-acetyl-2,3,6-trideoxy-3-phthalimido-hexofuranose 3 and 4. After acetylation at the anomeric center, coupling with silylated thymine resulted in three new nucleosides, with L-acosamine and L-ristosamine of furanose configuration as the carbohydrate moiety. The target compounds have been evaluated for their antiviral activity against HIV and HSV-1.