Merck
  • Effect of fumaric acid, its dimethylester, and topical antipsoriatic drugs on epidermal differentiation in the mouse tail model.
Effect of fumaric acid, its dimethylester, and topical antipsoriatic drugs on epidermal differentiation in the mouse tail model.
Skin pharmacology : the official journal of the Skin Pharmacology Society (1996-01-01)
B Sebök, T Szabados, M Kerényi, I Schneider, G Mahrle
ABSTRACT
Fumaric acid, fumaric acid dimethylester, and the dithranol derivative C4-lactone were studied in the mouse tail test to evaluate their effects on epidermal cell differentiation compared with other topical antipsoriatic drugs, such as betamethasone, calcipotriol, and dithranol. Mouse tails were treated for 2 weeks and longitudinal histological sections prepared of the tail skin. The length of the orthokeratotic regions (stratum granulosum) was measured on 10 sequential scales per tail and expressed as percentage of the full length of the scale. In addition, epidermal thickness was measured and the efficacy of the various compounds evaluated. In comparison to 2% salicylic acid ointment, all tested compounds except fumaric acid significantly (p < or = 0.05) increased the proportion of the orthokeratotic region. C4-lactone and calcipotriol were less effective than dithranol, fumaric acid dimethylester only moderately influenced cell differentiation, and betamethasone showed the least potent effect. Dithranol was the most potent substance inducing orthokeratosis without increasing epidermal thickness.
Sigma-Aldrich
Diethyl fumarate, 98%