Merck
  • An alternative synthesis of the NMDA antagonist CGS 19755 via free radical carbamoylation of ethyl isonicotinate.
An alternative synthesis of the NMDA antagonist CGS 19755 via free radical carbamoylation of ethyl isonicotinate.
Acta chemica Scandinavica (Copenhagen, Denmark : 1989) (1995-03-01)
I Martin, J Anvelt, L Vares, I Kühn, A Claesson
ABSTRACT
The NMDA antagonist CGS 19755 (cis-4-phosphonomethyl-2-piperidinecarboxylic acid) has been prepared by applying Minisci reaction conditions [formamide, hydrogen peroxide, iron(II) sulfate] to ethyl isonicotinate, reduction of the ester with sodium borohydride, alcoholysis of the 2-carboxamide, formation of 4-(diethylphosphonomethyl)-2-pyridinecarboxylate, hydrogenation of the pyridine nucleus, and acid hydrolysis. The overall, unoptimized yield was around 11%. The procedure employs cheap starting materials, is practical and avoids the use of toxic and hazardous cyanotrimethylsilane which is used in the published procedure.
Sigma-Aldrich
Ethyl isonicotinate, 98%