Merck
  • Schedule-dependency assessments of ribonucleoside diphosphate reductase inhibitors when used in combination with platinum compounds plus cyclophosphamide in the treatment of advanced L1210 leukemia.
Schedule-dependency assessments of ribonucleoside diphosphate reductase inhibitors when used in combination with platinum compounds plus cyclophosphamide in the treatment of advanced L1210 leukemia.
Cancer treatment reports (1979-03-01)
G R Gale, L M Atkins, S J Meischen, P Schwartz
PMID427825
ABSTRACT
Each of three ribonucleoside diphosphate reductase inhibitors was used as a third drug in combination with selected antitumor platinum (Pt) agents and cyclophosphamide (CY) in the treatment of advanced L1210 leukemia in C57BL/6 x DBA/2 mice. Each was synergistic with the various Pt plus CY combinations but the effect was highly schedule dependent. The collective cure rate was 68% when hydroxyurea (HU) was given as a single injection with Pt plus CY; the cure rate was 15% when HU was administered on a divided-dose schedule with Pt plus CY. The collective cure rate was 53% when guanazole was given as a single injection with Pt plus CY, but was only 8% when it was given on a divided-dose schedule with Pt plus CY. The effect of 4-methyl-5-amino-1-formylisoquinoline thiosemicarbazone, when used as a third drug with the various Pt plus CY regimens, was not schedule dependent as assessed by the collective cure rate. A therapeutic synergy between CY and each of the three ribonucleoside diphosphate reductase inhibitors was also observed.
Sigma-Aldrich
3,5-Diamino-1,2,4-triazole, 98%