Merck
  • Effects of monensin, pyromellitic diimide and 2-bromoethanesulfonic acid on rumen fermentation in vitro.
Effects of monensin, pyromellitic diimide and 2-bromoethanesulfonic acid on rumen fermentation in vitro.
Journal of animal science (1985-02-01)
S A Martin, J M Macy
ABSTRACT
The effects of monensin, 2-bromoethanesulfonic acid (2-BES) and pyromellitic diimide (diimide) on gas and volatile fatty acid (VFA) production by the rumen microbiota were compared in mixed culture. Oat hay, a hay-concentrate mixture (48% hay, 43% concentrate) and a soluble carbohydrate mixture were used as substrates for microbial growth. The highest concentrations of diimide (10 ppm) and 2-BES (30 microM) decreased methanogenesis by 97 and 76%, respectively, while H2 accumulation was increased 30- and 20-fold, respectively. The effect of monensin on methanogenesis was less dramatic as 10 ppm decreased CH4 accumulation 16% and H2 did not accumulate. Diimide and 2-BES decreased the acetate:propionate ratio with the hay (P less than .05) and soluble carbohydrate mixture (P less than .025). The numbers of saccharolytic, cellulolytic and methanogenic bacteria from sheep fed a diet with diimide (60 ppm) did not differ significantly from sheep fed a control diet. Semicontinuous culture incubations indicated that the mixed rumen microbial population could adapt and degrade diimide after 24 h of incubation.
Sigma-Aldrich
Pyromellitic diimide, 97%