Merck
  • Phase II open label, multi-center clinical trial of modulation of intermediate endpoint biomarkers by 1α-hydroxyvitamin D2 in patients with clinically localized prostate cancer and high grade pin.
Phase II open label, multi-center clinical trial of modulation of intermediate endpoint biomarkers by 1α-hydroxyvitamin D2 in patients with clinically localized prostate cancer and high grade pin.
The Prostate (2013-01-22)
Jason Gee, Howard Bailey, Kyungmann Kim, Jill Kolesar, Tom Havighurst, Kendra D Tutsch, William See, Michael B Cohen, Nick Street, Leon Levan, David Jarrard, George Wilding
ABSTRACT
Prostate cancer is the most common malignancy and second leading cause of cancer related deaths in American men supporting the study of prostate cancer chemoprevention. Major risk factors for this disease have been associated with low serum levels of vitamin D. Here, we evaluate the biologic activity of a less calcemic vitamin D analog 1α-hydroxyvitamin D2 [1α-OH-D2] (Bone Care International, Inc.) in patients with prostate cancer and high grade prostatic intraepithelial neoplasia (HG PIN). Patients with clinically organ-confined prostate cancer and HG PIN were randomized to 1α-OH-D2 versus placebo for 28 days prior to radical prostatectomy. Intermediate endpoint biomarkers included serum vitamin D metabolites, TGFß 1/2, free/total PSA, IGF-1, IGFBP-3, bFGF, and VEGF. Tissue endpoints included histology, MIB-1 and TUNEL staining, microvessel density and factor VIII staining, androgen receptor and PSA, vitamin D receptor expression and nuclear morphometry. The 1α-OH-D2 vitamin D analog was well tolerated and could be safely administered with good compliance and no evidence of hypercalcemia over 28 days. While serum vitamin D metabolite levels only slightly increased, evidence of biologic activity was observed with significant reductions in serum PTH levels. TGF-ß2 was the only biomarker significantly altered by vitamin D supplementation. Whether reduced TGF-ß2 levels in our study is an early indicator of response to vitamin D remains unclear. While further investigation of vitamin D may be warranted based on preclinical studies, results of the present trial do not appear to justify evaluation of 1α-OH-D2 in larger clinical prostate cancer prevention studies.
Sigma-Aldrich
Doxercalciferol, ≥98% (HPLC), solubility: >10 mg/mL in DMSO