Merck
  • Impact of pharmacogenetic markers of CYP2B6, clinical factors, and drug-drug interaction on efavirenz concentrations in HIV/tuberculosis-coinfected patients.
Impact of pharmacogenetic markers of CYP2B6, clinical factors, and drug-drug interaction on efavirenz concentrations in HIV/tuberculosis-coinfected patients.
Antimicrobial agents and chemotherapy (2012-12-21)
Weerawat Manosuthi, Chonlaphat Sukasem, Aroon Lueangniyomkul, Wiroj Mankatitham, Supeda Thongyen, Samruay Nilkamhang, Sukanya Manosuthi, Somnuek Sungkanuparph
ABSTRACT
Comprehensive information on the effects of cytochrome P450 2B6 (CYP2B6) polymorphisms, clinical factors, and drug-drug interactions on efavirenz concentrations in HIV/tuberculosis-coinfected (HIV/TB) patients is unavailable. A total of 139 HIV/TB adults, 101 of whom received a rifampin-containing anti-TB regimen, were prospectively enrolled to receive efavirenz (600 mg)/tenofovir/lamivudine. Nine single nucleotide polymorphisms (SNPs) within CYP2B6 were genotyped. Plasma efavirenz concentrations were measured at 12 weeks. The median (interquartile range [IQR]) efavirenz concentration was 2.3 (1.4 to 3.9) mg/liter. The SNPs (frequencies of heterozygous and homozygous mutants) were 64C>T (10% and 1%), 499C>G (0% and 0%), 516G>T (47% and 8%), 785A>G (54% and 10%), 1375A>G (0% and 0%), 1459C>T (3% and 0%), 3003C>T (44% and 27%), 18492T>C (39% and 6%), and 21563C>T (57% and 5%). The four most frequent CYP2B6 haplotypes identified were *1/*6 (41%), *1/*1 (35%), *1/*2 (7%), and *6/*6 (7%). The heterozygous/homozygous mutation associated with low efavirenz concentrations was 18492T>C (P < 0.001), and those associated with high efavirenz concentrations were 516G>T, 785A>G, and 21563C>T (all P < 0.05). Haplotype *1/*1 was associated with low efavirenz concentrations, and *6/*6, *1/*6, and *5/6 were associated with high efavirenz concentrations. As shown by multivariate analysis, low efavirenz concentrations were significantly associated with the *1/*1 haplotype (beta = -1.084, P = 0.027) and high body weight (beta = -0.076, P = 0.002). In conclusion, pharmacogenetic markers of CYP2B6 have the greatest impact with respect to inducing low plasma efavirenz concentrations in HIV/TB Thai patients.
Sigma-Aldrich
Pyrazinecarboxamide