Merck
  • Oxidation reactions of 2-thiouracil: a theoretical and pulse radiolysis study.
Oxidation reactions of 2-thiouracil: a theoretical and pulse radiolysis study.
The journal of physical chemistry. A (2012-10-16)
K P Prasanthkumar, C H Suresh, C T Aravindakumar
ABSTRACT
The reaction of hydroxyl radical ((•)OH) with the nucleic acid base analogue 2-thiouracil (1) has been studied by pulse radiolysis experiments and DFT. The generic intermediate radicals feasible for the (•)OH reactions with 1, namely, one electron oxidation product (1(•+)), (•)OH-adducts (3(•), 4(•), and 5(•)), and H-abstracted radicals (6(•) and 7(•)), were characterized by interpreting their electronic and structural properties along with calculated energetics and UV-vis spectra. Pulse radiolysis experiments showed that the transient formed in the reaction of (•)OH with 1 in water at pH 6.5 has λ(max) at 430 nm. A bimolecular rate constant, k(2) of 9.6 × 10(9) M(-1) s(-1), is determined for this reaction via competition kinetics with 2-propanol. The experiments suggested that the transient species could be a dimer radical cation 2(•+), formed by the reaction of 1 with the radical cation 1(•+). For this reaction, an equilibrium constant of 4.7 × 10(3) M(-1) was determined. The transient formed in the reaction of 1 with pulse radiolytically produced Br(2)(•-) at pH 6.5 as well as Cl(2)(•-) at pH 1 has also produced λ(max) at 430 nm and suggested the formation of 2(•+). The calculated UV-vis spectra of the transient species (1(•+), 3(•), 4(•), 5(•), 6(•), and 7(•)) showed no resemblance to the experimental spectra, while that of 2(•+) (λ(max) = 420 nm) agreed well with the experimental value and thus confirmed the formation of 2(•+). The 420 nm peak was due to σ → σ* electronic excitation centered on a 2-center-3-electron (2c-3e) sulfur-sulfur bond [-S∴S-]. 2(•+) is the first reported example of a dimer radical cation in a pyrimidine heterocyclic system. Further, 5-C and 6-C substituted (substituents are -F, -Cl, -NH(2), -N(CH(3))(2), -OCH(3), -CF(3), -CH(3), -CH(2)CH(3), n-propyl, phenyl, and benzyl) and 5,6-disubstituted 2-thiouracil systems have been characterized by DFT and found that the reaction (1 + 1(•+) → 2(•+)) is exergonic (1.12-13.63 kcal/mol) for many of them.
Sigma-Aldrich
2-Thiouracil, ≥99%
Pricing and availability is not currently available.